Assay Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001333.2 | NP_001001333.2 | 11 - 12 | 1627 | 87 | |
| NM_001146073.1 | NP_001139545.1 | 10 - 11 | 1432 | 87 | ||
| GenBank mRNA | AK154237.1 | - | 11 - 12 | 1601 | 87 | |
| BC069960.1 | - | 10 - 11 | 1384 | 5007884 | 87 | |
| FM204885.1 | - | 10 - 11 | 1164 | 87 | ||
| FM204886.1 | - | 10 - 11 | 1383 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexdc
Genomic Map