Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800005972.mm.1 |
NM_001001491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005972.mm.1 |
NM_001001491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005972.mm.1 |
TC0800000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005974.mm.1 |
NM_001001491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005974.mm.1 |
NM_001001491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005974.mm.1 |
TC0800000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005971.mm.1 |
NM_001001491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005971.mm.1 |
NM_001001491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005971.mm.1 |
TC0800000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005973.mm.1 |
NM_001001491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005973.mm.1 |
NM_001001491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005973.mm.1 |
TC0800000814.mm.1 |
MTA TC |