Assay Details
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167991.1 | NP_001161463.1 | 19 - 20 | 1931 | 64 | |
NM_133255.2 | NP_573518.2 | 19 - 20 | 1934 | 64 | ||
GenBank mRNA | AK030599.1 | - | 21 - 22 | 1950 | 64 | |
AK042048.1 | - | 19 - 20 | 1922 | 64 | ||
AK136069.1 | - | 20 - 21 | 1856 | 64 | ||
AK145768.1 | - | 19 - 20 | 1902 | 64 | ||
AK166954.1 | - | 19 - 20 | 1911 | 64 | ||
AK169073.1 | - | 19 - 20 | 1902 | 64 | ||
BC002226.1 | - | 11 - 12 | 1143 | 64 | ||
BC019486.1 | - | 19 - 20 | 1910 | 4211990 | 64 | |
BC026609.1 | - | 11 - 12 | 1094 | 64 | ||
BC029967.1 | - | 19 - 20 | 1875 | 64 | ||
BC055881.1 | - | 19 - 20 | 1877 | 3590336 | 64 | |
BC080744.1 | - | 19 - 20 | 1924 | 30471369 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
