Assay Details
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167991.1 | NP_001161463.1 | 20 - 21 | 2039 | 58 | |
NM_133255.2 | NP_573518.2 | 20 - 21 | 2042 | 58 | ||
GenBank mRNA | AK030599.1 | - | 22 - 23 | 2058 | 58 | |
AK042048.1 | - | 20 - 21 | 2030 | 58 | ||
AK136069.1 | - | 21 - 22 | 1964 | 58 | ||
AK145768.1 | - | 20 - 21 | 2010 | 58 | ||
AK166954.1 | - | 20 - 21 | 2019 | 58 | ||
AK169073.1 | - | 20 - 21 | 2010 | 58 | ||
BC002226.1 | - | 12 - 13 | 1251 | 58 | ||
BC019486.1 | - | 20 - 21 | 2018 | 4211990 | 58 | |
BC026609.1 | - | 12 - 13 | 1202 | 58 | ||
BC029967.1 | - | 20 - 21 | 1983 | 58 | ||
BC055881.1 | - | 20 - 21 | 1985 | 3590336 | 58 | |
BC080744.1 | - | 20 - 21 | 2032 | 30471369 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
