Assay Details
Gene Symbol: Prss46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183103.2 | NP_898926.2 | 2 - 3 | 307 | 60 | |
GenBank mRNA | AK007173.1 | - | 2 - 3 | 306 | 60 | |
BC087878.1 | - | 1 - 2 | 264 | 30284660 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
