| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_001146059.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_001146059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_001146060.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_001146060.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_146228.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
NM_146228.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900012378.mm.1 |
TC0900001475.mm.1 |
MTA TC |