Assay Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019726.3 | NP_062700.2 | 8 - 9 | 948 | 72 | |
| GenBank mRNA | AF153696.1 | - | 9 - 10 | 888 | 72 | |
| BC011317.1 | - | 8 - 9 | 948 | 3979292 | 72 | |
| BC138879.1 | - | 8 - 9 | 904 | 8861901 | 72 | |
| BC138880.1 | - | 8 - 9 | 905 | 8861902 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gps2
Genomic Map