Assay Details
| Gene Symbol: Eif5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166592.1 | NP_001160064.1 | 1 - 2 | 150 | 95 | |
| GenBank mRNA | AY129329.1 | - | 1 - 2 | 143 | 95 | |
| CX237346.1 | - | 1 - 2 | 139 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AY129329.1,NM_001166592.1 transcripts.
This assay is in the 5'UTR region of the AY129329.1,NM_001166592.1 transcripts.
Set Membership: |
|

Eif5a
Genomic Map