Assay Details
Gene Symbol: Pelp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029231.4 | NP_083507.3 | 5 - 6 | 662 | 85 | |
GenBank mRNA | AK005027.1 | - | 5 - 6 | 660 | 85 | |
AK128966.1 | - | 5 - 6 | 662 | 85 | ||
BC090620.1 | - | 5 - 6 | 656 | 30699968 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
