Assay Details
Gene Symbol: Pelp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029231.4 | NP_083507.3 | 7 - 8 | 870 | 80 | |
GenBank mRNA | AK005027.1 | - | 7 - 8 | 868 | 80 | |
AK128966.1 | - | 7 - 8 | 870 | 80 | ||
BC057987.1 | - | 1 - 2 | 98 | 80 | ||
BC090620.1 | - | 7 - 8 | 864 | 30699968 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
