Assay Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007440.4 | NP_031466.2 | 5 - 6 | 714 | 92 | |
| GenBank mRNA | AK036898.1 | - | 5 - 6 | 712 | 92 | |
| AK036960.1 | - | 5 - 6 | 670 | 92 | ||
| AK079232.1 | - | 5 - 6 | 715 | 92 | ||
| AK087724.1 | - | 5 - 6 | 715 | 92 | ||
| AK172216.1 | - | 5 - 6 | 712 | 92 | ||
| BC152329.1 | - | 5 - 6 | 719 | 40141477 | 92 | |
| BC152403.1 | - | 5 - 6 | 719 | 40141482 | 92 | |
| S80446.1 | - | 5 - 6 | 726 | 92 | ||
| U04334.1 | - | 5 - 6 | 645 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alox12
Genomic Map