Assay Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007440.4 | NP_031466.2 | 9 - 10 | 1316 | 86 | |
| GenBank mRNA | AK036898.1 | - | 9 - 10 | 1314 | 86 | |
| AK036960.1 | - | 9 - 10 | 1272 | 86 | ||
| AK087724.1 | - | 9 - 10 | 1317 | 86 | ||
| AK172216.1 | - | 9 - 10 | 1314 | 86 | ||
| AK172488.1 | - | 9 - 10 | 1317 | 86 | ||
| BC152329.1 | - | 9 - 10 | 1321 | 40141477 | 86 | |
| BC152403.1 | - | 9 - 10 | 1321 | 40141482 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alox12
Genomic Map