Assay Details
| Gene Symbol: Mlycd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019966.2 | NP_064350.2 | 1 - 2 | 581 | 76 | |
| GenBank mRNA | AK051564.1 | - | 1 - 2 | 531 | 76 | |
| AK082479.1 | - | 1 - 2 | 556 | 76 | ||
| AK171095.1 | - | 1 - 2 | 575 | 76 | ||
| BC004764.1 | - | 1 - 2 | 581 | 3155699 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlycd
Genomic Map