Assay Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291141.1 | NP_001278070.1 | 6 - 7 | 1047 | 139 | |
NM_001291142.1 | NP_001278071.1 | 6 - 7 | 1056 | 139 | ||
NM_001291143.1 | NP_001278072.1 | 8 - 9 | 1247 | 139 | ||
NM_019400.3 | NP_062273.2 | 7 - 8 | 1176 | 139 | ||
GenBank mRNA | AB001750.1 | - | 8 - 9 | 1083 | 139 | |
AF248489.1 | - | 6 - 7 | 842 | 139 | ||
AF248490.1 | - | 6 - 7 | 833 | 139 | ||
AK134587.1 | - | 7 - 8 | 1176 | 139 | ||
AK137236.1 | - | 7 - 8 | 1141 | 139 | ||
BC060166.1 | - | 7 - 8 | 1155 | 139 | ||
BC080274.1 | - | 7 - 8 | 1149 | 139 | ||
BC129853.1 | - | 7 - 8 | 995 | 40109688 | 139 | |
BC129854.1 | - | 7 - 8 | 995 | 40109689 | 139 | |
D86066.1 | - | 7 - 8 | 1012 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
