Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028843.mm.1 |
NM_001083331.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028843.mm.1 |
NM_001276406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028843.mm.1 |
NM_172394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028843.mm.1 |
NR_075087.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028843.mm.1 |
TC1100003117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028846.mm.1 |
NM_001083331.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028846.mm.1 |
NM_001276406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028846.mm.1 |
NM_172394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028846.mm.1 |
NR_075087.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028846.mm.1 |
TC1100003117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028845.mm.1 |
NM_001083331.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028845.mm.1 |
NM_001276406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028845.mm.1 |
NM_172394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028845.mm.1 |
NR_075087.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028845.mm.1 |
TC1100003117.mm.1 |
MTA TC |