Assay Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172394.3 | NP_765982.2 | 11 - 12 | 1664 | 130 | |
| NR_075087.1 | 10 - 11 | 2123 | 130 | |||
| GenBank mRNA | AK085776.1 | - | 11 - 12 | 1663 | 130 | |
| AK139738.1 | - | 10 - 11 | 2031 | 130 | ||
| AK160619.1 | - | 11 - 12 | 1665 | 130 | ||
| BC032929.1 | - | 11 - 12 | 1645 | 1379673 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup88
Genomic Map