Assay Details
| Gene Symbol: Spns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029932.3 | NP_084208.1 | 4 - 5 | 605 | 77 | |
| GenBank mRNA | AK020683.1 | - | 4 - 5 | 606 | 77 | |
| AK041376.1 | - | 3 - 4 | 595 | 77 | ||
| AK156479.1 | - | 4 - 5 | 869 | 77 | ||
| AK156781.1 | - | 4 - 5 | 598 | 76 | ||
| BC150778.1 | - | 4 - 5 | 588 | 9087689 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spns3
Genomic Map