Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200041528.mm.1 |
NM_028666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041528.mm.1 |
TC0200005510.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041525.mm.1 |
NM_001289150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041525.mm.1 |
NM_001289151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041525.mm.1 |
NM_028666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041525.mm.1 |
NM_146127.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041525.mm.1 |
TC0200005510.mm.1 |
MTA TC |