Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 5 - 6 | 682 | 88 | |
| GenBank mRNA | AK010967.1 | - | 5 - 6 | 511 | 88 | |
| AK137542.1 | - | 5 - 6 | 578 | 88 | ||
| AK157181.1 | - | 5 - 6 | 651 | 88 | ||
| AK157833.1 | - | 5 - 6 | 540 | 88 | ||
| AK157861.1 | - | 5 - 6 | 552 | 88 | ||
| AK158143.1 | - | 5 - 6 | 683 | 88 | ||
| AK158184.1 | - | 5 - 6 | 648 | 88 | ||
| AK158243.1 | - | 5 - 6 | 552 | 88 | ||
| AK158453.1 | - | 5 - 6 | 527 | 88 | ||
| AK162616.1 | - | 5 - 6 | 527 | 88 | ||
| AK165195.1 | - | 5 - 6 | 539 | 88 | ||
| AK165267.1 | - | 5 - 6 | 539 | 88 | ||
| AK171662.1 | - | 578 | 88 | |||
| AK171808.1 | - | 5 - 6 | 516 | 88 | ||
| AK171845.1 | - | 578 | 88 | |||
| AK172103.1 | - | 5 - 6 | 541 | 86 | ||
| AK172316.1 | - | 5 - 6 | 527 | 88 | ||
| AK172395.1 | - | 5 - 6 | 578 | 88 | ||
| AK172456.1 | - | 5 - 6 | 564 | 88 | ||
| AK172569.1 | - | 5 - 6 | 527 | 88 | ||
| AY296129.1 | - | 5 - 6 | 515 | 88 | ||
| BC052419.1 | - | 5 - 6 | 522 | 5702599 | 88 | |
| BC053429.1 | - | 5 - 6 | 513 | 6506270 | 88 | |
| M29379.1 | - | 5 - 6 | 509 | 88 | ||
| X02677.1 | - | 5 - 6 | 515 | 88 | ||
| X03917.1 | - | 3 - 4 | 359 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK165267.1 transcripts.
Set Membership: |
|

Slc4a1
Genomic Map