Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 9 - 10 | 1213 | 63 | |
| GenBank mRNA | AK137542.1 | - | 9 - 10 | 1109 | 63 | |
| AK157181.1 | - | 9 - 10 | 1182 | 63 | ||
| AK157833.1 | - | 9 - 10 | 1071 | 63 | ||
| AK157861.1 | - | 9 - 10 | 1083 | 63 | ||
| AK158143.1 | - | 9 - 10 | 1214 | 63 | ||
| AK158184.1 | - | 9 - 10 | 1179 | 63 | ||
| AK158243.1 | - | 9 - 10 | 63 | |||
| AK158453.1 | - | 9 - 10 | 1058 | 63 | ||
| AK162616.1 | - | 9 - 10 | 1058 | 63 | ||
| AK165195.1 | - | 9 - 10 | 1070 | 63 | ||
| AK165267.1 | - | 9 - 10 | 1074 | 63 | ||
| AK171662.1 | - | 1109 | 63 | |||
| AK171808.1 | - | 9 - 10 | 1047 | 63 | ||
| AK171845.1 | - | 1109 | 63 | |||
| AK172103.1 | - | 9 - 10 | 1070 | 63 | ||
| AK172316.1 | - | 9 - 10 | 1058 | 63 | ||
| AK172395.1 | - | 9 - 10 | 1109 | 63 | ||
| AK172456.1 | - | 9 - 10 | 1093 | 63 | ||
| AK172569.1 | - | 9 - 10 | 1058 | 63 | ||
| AY296129.1 | - | 9 - 10 | 1046 | 63 | ||
| BC052419.1 | - | 9 - 10 | 1053 | 5702599 | 63 | |
| BC053429.1 | - | 9 - 10 | 1044 | 6506270 | 63 | |
| M29379.1 | - | 9 - 10 | 1040 | 63 | ||
| X02677.1 | - | 9 - 10 | 1046 | 63 | ||
| X03917.1 | - | 7 - 8 | 890 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK165267.1 transcripts.
Set Membership: |
|

Slc4a1
Genomic Map