Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 13 - 14 | 1972 | 86 | |
| GenBank mRNA | AK137542.1 | - | 13 - 14 | 1868 | 86 | |
| AK157181.1 | - | 13 - 14 | 1941 | 86 | ||
| AK157833.1 | - | 13 - 14 | 1830 | 86 | ||
| AK157861.1 | - | 13 - 14 | 1842 | 86 | ||
| AK158143.1 | - | 13 - 14 | 1973 | 86 | ||
| AK158184.1 | - | 13 - 14 | 1938 | 86 | ||
| AK158243.1 | - | 13 - 14 | 1842 | 86 | ||
| AK158453.1 | - | 13 - 14 | 1817 | 86 | ||
| AK162616.1 | - | 13 - 14 | 1817 | 86 | ||
| AK165195.1 | - | 13 - 14 | 1829 | 86 | ||
| AK165267.1 | - | 13 - 14 | 1836 | 86 | ||
| AK171662.1 | - | 1868 | 86 | |||
| AK171808.1 | - | 13 - 14 | 1806 | 86 | ||
| AK171845.1 | - | 1868 | 86 | |||
| AK172103.1 | - | 13 - 14 | 1829 | 86 | ||
| AK172316.1 | - | 13 - 14 | 1817 | 86 | ||
| AK172395.1 | - | 13 - 14 | 1868 | 86 | ||
| AK172456.1 | - | 13 - 14 | 1852 | 86 | ||
| AK172569.1 | - | 13 - 14 | 1817 | 86 | ||
| AY296129.1 | - | 13 - 14 | 1805 | 86 | ||
| BC052419.1 | - | 13 - 14 | 1812 | 5702599 | 86 | |
| BC053429.1 | - | 13 - 14 | 1803 | 6506270 | 86 | |
| M29379.1 | - | 13 - 14 | 1799 | 86 | ||
| X02677.1 | - | 13 - 14 | 1805 | 86 | ||
| X03917.1 | - | 11 - 12 | 1649 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a1
Genomic Map