Assay Details
| Gene Symbol: Srr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163311.1 | NP_001156783.1 | 2 - 3 | 350 | 92 | |
| NM_013761.4 | NP_038789.1 | 3 - 4 | 576 | 92 | ||
| GenBank mRNA | AF148321.1 | - | 2 - 3 | 231 | 92 | |
| AK017235.1 | - | 3 - 4 | 456 | 92 | ||
| AK028034.1 | - | 2 - 3 | 233 | 92 | ||
| AK031687.1 | - | 3 - 4 | 448 | 92 | ||
| AK043738.1 | - | 3 - 4 | 482 | 92 | ||
| AK043819.1 | - | 2 - 3 | 256 | 92 | ||
| AK080830.1 | - | 3 - 4 | 93 | |||
| AK157122.1 | - | 2 - 3 | 501 | 92 | ||
| AK170096.1 | - | 3 - 4 | 462 | 92 | ||
| AK170471.1 | - | 3 - 4 | 462 | 92 | ||
| BC011164.1 | - | 3 - 4 | 453 | 4195695 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srr
Genomic Map