Assay Details
| Gene Symbol: Gosr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016810.3 | NP_058090.2 | 2 - 3 | 154 | 67 | |
| GenBank mRNA | AF079901.1 | - | 2 - 3 | 177 | 67 | |
| AK133836.1 | - | 2 - 3 | 164 | 67 | ||
| AK134515.1 | - | 2 - 3 | 157 | 67 | ||
| AK147799.1 | - | 2 - 3 | 178 | 67 | ||
| AK154972.1 | - | 2 - 3 | 180 | 67 | ||
| AK161386.1 | - | 2 - 3 | 168 | 67 | ||
| BC008542.1 | - | 2 - 3 | 154 | 3495882 | 67 | |
| BC051661.1 | - | 2 - 3 | 30007684 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gosr1
Genomic Map