Assay Details
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112705.2 | NP_001106176.1 | 13 - 14 | 1716 | 103 | |
NM_001294329.1 | NP_001281258.1 | 11 - 12 | 1576 | 103 | ||
NM_001294330.1 | NP_001281259.1 | 11 - 12 | 1524 | 103 | ||
NM_001294331.1 | NP_001281260.1 | 13 - 14 | 1729 | 103 | ||
NM_001294334.1 | NP_001281263.1 | 13 - 14 | 1716 | 103 | ||
GenBank mRNA | AF045252.1 | - | 13 - 14 | 1735 | 103 | |
AF045253.1 | - | 12 - 13 | 1163 | 103 | ||
AF045254.1 | - | 8 - 9 | 877 | 103 | ||
AK014829.1 | - | 13 - 14 | 1354 | 103 | ||
AK089925.1 | - | 11 - 12 | 1104 | 103 | ||
AK165667.1 | - | 13 - 14 | 1719 | 103 | ||
AX775743.1 | - | 7 - 8 | 741 | 103 | ||
BC066198.1 | - | 11 - 12 | 1069 | 103 | ||
U56909.1 | - | 4 - 5 | 414 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
