Assay Details
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112705.2 | NP_001106176.1 | 9 - 10 | 1244 | 135 | |
NM_001294329.1 | NP_001281258.1 | 7 - 8 | 1104 | 135 | ||
NM_001294331.1 | NP_001281260.1 | 9 - 10 | 1257 | 135 | ||
NM_001294334.1 | NP_001281263.1 | 9 - 10 | 1244 | 135 | ||
GenBank mRNA | AF045252.1 | - | 9 - 10 | 1263 | 135 | |
AF045253.1 | - | 8 - 9 | 691 | 135 | ||
AF045254.1 | - | 4 - 5 | 405 | 135 | ||
AK014829.1 | - | 9 - 10 | 882 | 135 | ||
AK089925.1 | - | 7 - 8 | 632 | 135 | ||
AK165667.1 | - | 9 - 10 | 1247 | 135 | ||
AX775743.1 | - | 3 - 4 | 269 | 135 | ||
CD554252.1 | - | 3 - 4 | 368 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
