Assay Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033327.2 | NP_201584.2 | 4 - 5 | 3877 | 67 | |
| GenBank mRNA | AF188609.1 | - | 2 - 3 | 3246 | 67 | |
| AK122365.1 | - | 4 - 5 | 3843 | 67 | ||
| AK133497.1 | - | 2 - 3 | 3305 | 67 | ||
| AY147407.1 | - | 3 - 4 | 3482 | 67 | ||
| AY256893.1 | - | 4 - 5 | 3776 | 67 | ||
| BC059234.1 | - | 2 - 3 | 3688 | 67 | ||
| BC079586.1 | - | 4 - 5 | 3563 | 67 | ||
| BC139028.1 | - | 4 - 5 | 3623 | 8862050 | 67 | |
| BC139030.1 | - | 4 - 5 | 3623 | 8862052 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp423
Genomic Map