Assay Details
| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016905.2 | NP_058601.2 | 1 - 2 | 207 | 74 | |
| GenBank mRNA | AB027012.1 | - | 1 - 2 | 163 | 74 | |
| AK010340.1 | - | 1 - 2 | 204 | 74 | ||
| AK011103.1 | - | 1 - 2 | 208 | 74 | ||
| AK045694.1 | - | 1 - 2 | 181 | 74 | ||
| AK088151.1 | - | 1 - 2 | 208 | 74 | ||
| AK168716.1 | - | 1 - 2 | 167 | 74 | ||
| BC016602.1 | - | 1 - 2 | 207 | 3490224 | 74 | |
| BC050151.1 | - | 1 - 2 | 178 | 30024030 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galk1
Genomic Map