Assay Details
| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016905.2 | NP_058601.2 | 3 - 4 | 511 | 60 | |
| GenBank mRNA | AB027012.1 | - | 3 - 4 | 467 | 60 | |
| AK010340.1 | - | 3 - 4 | 508 | 60 | ||
| AK011103.1 | - | 3 - 4 | 512 | 60 | ||
| AK045694.1 | - | 3 - 4 | 485 | 60 | ||
| AK088151.1 | - | 3 - 4 | 512 | 60 | ||
| AK168716.1 | - | 3 - 4 | 471 | 60 | ||
| BC016602.1 | - | 3 - 4 | 511 | 3490224 | 60 | |
| BC050151.1 | - | 3 - 4 | 482 | 30024030 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galk1
Genomic Map