Assay Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012030.2 | NP_036160.1 | 4 - 5 | 974 | 108 | |
| GenBank mRNA | AK153558.1 | - | 4 - 5 | 974 | 108 | |
| AK168894.1 | - | 4 - 5 | 975 | 108 | ||
| AK177064.1 | - | 2 - 3 | 154 | 109 | ||
| AK188344.1 | - | 2 - 3 | 154 | 108 | ||
| AK205456.1 | - | 2 - 3 | 154 | 108 | ||
| BC085141.1 | - | 4 - 5 | 959 | 6413140 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r1
Genomic Map