Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100039099.mm.1 |
NM_009180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039099.mm.1 |
NM_009180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039099.mm.1 |
TC1100004124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039102.mm.1 |
NM_009180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039102.mm.1 |
NM_009180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039102.mm.1 |
TC1100004124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039103.mm.1 |
NM_009180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039103.mm.1 |
NM_009180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039103.mm.1 |
TC1100004124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039101.mm.1 |
NM_009180.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039101.mm.1 |
NM_009180.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039101.mm.1 |
TC1100004124.mm.1 |
MTA TC |