Assay Details
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009180.3 | NP_033206.2 | 6 - 7 | 873 | 65 | |
GenBank mRNA | AK004613.1 | - | 6 - 7 | 874 | 65 | |
AK136381.1 | - | 876 | 65 | |||
BC003276.1 | - | 6 - 7 | 1775 | 65 | ||
BC010208.1 | - | 825 | 65 | |||
X93999.1 | - | 6 - 7 | 837 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
