Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 4 - 5 | 281 | 86 | |
| GenBank mRNA | AB039948.1 | - | 2 - 3 | 102 | 86 | |
| AK013520.1 | - | 4 - 5 | 280 | 86 | ||
| AK044296.1 | - | 4 - 5 | 273 | 86 | ||
| AK158225.1 | - | 4 - 5 | 283 | 86 | ||
| BC126874.1 | - | 2 - 3 | 95 | 40044848 | 86 | |
| BC126875.1 | - | 2 - 3 | 95 | 40044849 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB039948.1, BC126874.1, BC126875.1 transcripts.
Set Membership: |
|

Necab3
Genomic Map