Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 10 - 11 | 879 | 84 | |
| GenBank mRNA | AB039948.1 | - | 8 - 9 | 700 | 84 | |
| AK013520.1 | - | 9 - 10 | 806 | 84 | ||
| AK044296.1 | - | 10 - 11 | 871 | 84 | ||
| AK158225.1 | - | 10 - 11 | 881 | 84 | ||
| BC126874.1 | - | 8 - 9 | 693 | 40044848 | 84 | |
| BC126875.1 | - | 8 - 9 | 693 | 40044849 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Necab3
Genomic Map