Assay Details
Gene Symbol: Sgsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197943.2 | NP_922934.2 | 7 - 8 | 1003 | 60 | |
GenBank mRNA | AK122273.1 | - | 7 - 8 | 925 | 60 | |
AK208574.1 | - | 1 - 2 | 67 | 60 | ||
BC058414.1 | - | 7 - 8 | 994 | 6822416 | 60 | |
BC060163.1 | - | 7 - 8 | 994 | 6822416 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
