| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200042055.mm.1 |
NM_001004721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042055.mm.1 |
NM_001004721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042055.mm.1 |
TC0200004958.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042057.mm.1 |
NM_001004721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042057.mm.1 |
NM_001004721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042057.mm.1 |
TC0200004958.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042056.mm.1 |
NM_001004721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042056.mm.1 |
NM_001004721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042056.mm.1 |
TC0200004958.mm.1 |
MTA TC |