Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200042192.mm.1 |
NM_001163452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042192.mm.1 |
NM_001163452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042192.mm.1 |
NM_019828.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042192.mm.1 |
NM_019828.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042192.mm.1 |
TC0200004967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042195.mm.1 |
NM_001163452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042195.mm.1 |
NM_001163452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042195.mm.1 |
NM_019828.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042195.mm.1 |
NM_019828.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042195.mm.1 |
TC0200004967.mm.1 |
MTA TC |