Assay Details
| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291105.1 | NP_001278034.1 | 3 - 4 | 669 | 71 | |
| NM_007891.5 | NP_031917.1 | 3 - 4 | 713 | 71 | ||
| GenBank mRNA | AF483514.1 | - | 3 - 4 | 555 | 71 | |
| AF483515.1 | - | 3 - 4 | 555 | 71 | ||
| AK017841.1 | - | 3 - 4 | 661 | 71 | ||
| AK153968.1 | - | 3 - 4 | 678 | 71 | ||
| AK211069.1 | - | 1 - 2 | 66 | 71 | ||
| AK220226.1 | - | 3 - 4 | 410 | 71 | ||
| BC052160.1 | - | 3 - 4 | 655 | 6330425 | 71 | |
| L21973.1 | - | 3 - 4 | 673 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E2f1
Genomic Map