Assay Details
| Gene Symbol: Soga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164663.1 | NP_001158135.1 | 15 | 102 | ||
| GenBank mRNA | AK052498.1 | - | 1 | 102 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0200005011.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0200005011.mm.1| Set Membership: |  Amplicon length greater than or equal to 101  Detect Genomic DNA  Recommended Assay  Mus musculus  1700  Both primers and probe map within a single exon | 
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Soga1
Soga1 Genomic Map
Genomic Map Back To Top
 Back To Top