Assay Details
| Gene Symbol: Spag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139151.4 | NP_631890.3 | 4 - 5 | 694 | 67 | |
| GenBank mRNA | AB041554.1 | - | 2 - 3 | 1273 | 67 | |
| AK028080.1 | - | 4 - 5 | 663 | 67 | ||
| AK043812.1 | - | 4 - 5 | 662 | 67 | ||
| AK220494.1 | - | 1670 | 67 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Spag4
Genomic Map