Assay Details
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139151.4 | NP_631890.3 | 11 - 12 | 1318 | 135 | |
GenBank mRNA | AB041554.1 | - | 9 - 10 | 1897 | 135 | |
AK220494.1 | - | 2591 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769252 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769255 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769251 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769253 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513992 | AK028080.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513992 | AK028080.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765774 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107613 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124388 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513992 | AK028080.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513992 | AK028080.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769249 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124388 | - | - |
Whole Mouse Genome G4122A | A_51_P513991 | AK028080.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513992 | AK028080.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513992 | AK028080.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125423 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101990 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124388 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01458303 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769252 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769251 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769255 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01743644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01893686 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01176401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01287980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01287994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01515601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01835446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01515598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01287982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01511307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21769247 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10477792 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477796 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477807 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477798 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477797 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477793 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477806 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477794 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477799 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477800 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477795 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477804 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477801 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477802 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452407_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10477791 | - | - |
Mouse Genome 430 2.0 Array | 1452407_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452407_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245865_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245865_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200019143.mm.1 | NM_139151.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200019143.mm.1 | NM_139151.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200019143.mm.1 | TC0200002383.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200019142.mm.1 | NM_139151.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200019142.mm.1 | NM_139151.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200019142.mm.1 | TC0200002383.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452407_PM_at | - | - |
Mouse Expression Set 430 | 1452407_at | - | - |
Murine Genome U74v2 Set | 167850_PM_r_at | - | - |
Murine Genome U74v2 Set | 167850_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1452407_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452407_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452407_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452407_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 107040575 | - | - |
MouseWG-6 v2 Expression BeadChip | 2680373 | - | - |
MouseWG-6 v2 Expression BeadChip | 360128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2680373 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 360128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 107040575 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002680373 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000360128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243979 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659447 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2659447 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243979 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2504368 | - | - |
mouse-6 v1.1 (Illumina) | scl00245865.2_96-S | - | - |
mouse-6 v1.1 (Illumina) | scl20038.8.1_45-S | - | - |
mouse-6 v1.1 (Illumina) | scl0003160.1_38-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 360128 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 107040575 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2680373 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003160.1_38-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00245865.2_96-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107040575 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680373 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360128 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2659447 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2975391 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1243979 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2975391 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2504368 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243979 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2659447 | - | - |
