Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200042352.mm.1 |
NM_001145351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042352.mm.1 |
NM_001145351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042352.mm.1 |
NM_001145352.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042352.mm.1 |
NM_001145352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042352.mm.1 |
TC0200004979.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042350.mm.1 |
NM_001145351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042350.mm.1 |
NM_001145351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042350.mm.1 |
NM_001145352.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042350.mm.1 |
NM_001145352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042350.mm.1 |
TC0200004979.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042353.mm.1 |
NM_001145351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042353.mm.1 |
NM_001145351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042353.mm.1 |
NM_001145352.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042353.mm.1 |
NM_001145352.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042353.mm.1 |
TC0200004979.mm.1 |
MTA TC |