Assay Details
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291190.1 | NP_001278119.1 | 13 - 14 | 2341 | 153 | |
NM_177710.4 | NP_808378.2 | 14 - 15 | 2205 | 153 | ||
GenBank mRNA | AB099288.1 | - | 14 - 15 | 2205 | 153 | |
AK155232.1 | - | 13 - 14 | 2332 | 153 | ||
AK173243.1 | - | 3 - 4 | 1069 | 153 | ||
BC141392.1 | - | 14 - 15 | 2199 | 9056005 | 153 | |
BC141393.1 | - | 14 - 15 | 2199 | 9056006 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
