Assay Details
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291190.1 | NP_001278119.1 | 5 - 6 | 633 | 95 | |
NM_177710.4 | NP_808378.2 | 6 - 7 | 497 | 95 | ||
GenBank mRNA | AB099288.1 | - | 6 - 7 | 497 | 95 | |
AK155232.1 | - | 5 - 6 | 624 | 95 | ||
AK170142.1 | - | 5 - 6 | 630 | 95 | ||
BC141392.1 | - | 6 - 7 | 491 | 9056005 | 95 | |
BC141393.1 | - | 6 - 7 | 491 | 9056006 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
