Assay Details
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291190.1 | NP_001278119.1 | 6 - 7 | 698 | 114 | |
NM_177710.4 | NP_808378.2 | 7 - 8 | 562 | 114 | ||
GenBank mRNA | AB099288.1 | - | 7 - 8 | 562 | 114 | |
AK155232.1 | - | 6 - 7 | 689 | 114 | ||
AK170142.1 | - | 6 - 7 | 695 | 114 | ||
BC141392.1 | - | 7 - 8 | 556 | 9056005 | 114 | |
BC141393.1 | - | 7 - 8 | 556 | 9056006 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
