Assay Details
| Gene Symbol: Slc25a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013770.2 | NP_038798.2 | 3 - 4 | 471 | 98 | |
| GenBank mRNA | AF188712.1 | - | 3 - 4 | 442 | 98 | |
| AK008038.1 | - | 3 - 4 | 441 | 98 | ||
| AK050282.1 | - | 3 - 4 | 425 | 98 | ||
| AK083831.1 | - | 3 - 4 | 449 | 98 | ||
| AK151645.1 | - | 3 - 4 | 471 | 98 | ||
| BC003222.1 | - | 3 - 4 | 389 | 3482316 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a10
Genomic Map