Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026428.2 | NP_080704.2 | 3 - 4 | 354 | 76 | |
| GenBank mRNA | AK002803.1 | - | 3 - 4 | 324 | 76 | |
| AK004023.1 | - | 3 - 4 | 353 | 76 | ||
| AK007627.1 | - | 3 - 4 | 366 | 76 | ||
| AK153521.1 | - | 3 - 4 | 335 | 76 | ||
| AK161732.1 | - | 310 | 76 | |||
| AK178061.1 | - | 1 - 2 | 158 | 76 | ||
| BC012247.1 | - | 3 - 4 | 343 | 4189635 | 76 | |
| D89656.1 | - | 3 - 4 | 302 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcxr
Genomic Map