Assay Details
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024229.2 | NP_077191.2 | 13 - 14 | 1149 | 67 | |
GenBank mRNA | AK140458.1 | - | 13 - 14 | 1189 | 67 | |
AK144544.1 | - | 13 - 14 | 1206 | 67 | ||
AK166319.1 | - | 12 - 13 | 1124 | 67 | ||
BC003473.1 | - | 13 - 14 | 1155 | 2650077 | 67 | |
BC008276.1 | - | 13 - 14 | 1149 | 3707732 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185660 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185661 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185659 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112230 | BC003473.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112230 | BC008276.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112230 | AK140458.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112230 | AK144544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112230 | AK166319.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228143 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344632 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | AK031999.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | AK077642.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | AK131658.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | BC016084.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | BC027819.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | BC031181.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898317 | NM_001001181.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94502 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173156 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112230 | BC003473.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112230 | BC008276.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112230 | AK140458.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112230 | AK144544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112230 | AK166319.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432504 | BC003473.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432504 | BC008276.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432504 | NM_024229.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P142154 | BC003473.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P142154 | BC008276.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P142154 | AK140458.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P142154 | NM_024229.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P142154 | AK004054.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185645 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185611 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185660 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185653 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185658 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02446770 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02446775 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173156 | - | - |
Whole Mouse Genome G4122A | A_52_P142154 | BC003473.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P142154 | BC008276.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P142154 | AK140458.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P142154 | NM_024229.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P142154 | AK004054.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432504 | BC003473.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432504 | BC008276.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P432504 | NM_024229.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126947 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173156 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09612757 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09518142 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09518146 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185658 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185661 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185660 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09120660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09120653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09248780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09588015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09202799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09518142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09248785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28185620 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10393856 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393864 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393865 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393854 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393861 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393860 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393858 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393863 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393855 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393862 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393857 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10393853 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420493_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10393851 | - | - |
Mouse Genome 430 2.0 Array | 1420493_a_at | - | - |
Mouse Genome 430 2.0 Array | 1420493_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68671_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68671_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100040254.mm.1 | NM_024229.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040254.mm.1 | NM_024229.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040254.mm.1 | TC1100004220.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100040252.mm.1 | NM_024229.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040252.mm.1 | NM_024229.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040252.mm.1 | TC1100004220.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100040253.mm.1 | NM_024229.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040253.mm.1 | NM_024229.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100040253.mm.1 | TC1100004220.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420493_a_at | - | - |
Mouse Expression Set 430 | 1420493_PM_a_at | - | - |
Murine Genome U74v2 Set | 103914_at | - | - |
Murine Genome U74v2 Set | 103914_PM_at | - | - |
Murine Genome U74v2 Set | 168071_PM_i_at | - | - |
Murine Genome U74v2 Set | 139034_PM_at | - | - |
Murine Genome U74v2 Set | 168071_i_at | - | - |
Murine Genome U74v2 Set | 139034_at | - | - |
Mouse Genome 430A 2.0 Array | 1420493_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1420493_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420493_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420493_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2630092 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4590278 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002630092 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004590278 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694038 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597485 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2597485 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2694038 | - | - |
mouse-6 v1.1 (Illumina) | scl0001280.1_1-S | - | - |
mouse-6 v1.1 (Illumina) | scl39229.10.14_5-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2630092 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4590278 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001280.1_1-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39229.10.14_5-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630092 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590278 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2939303 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2597485 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2694038 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2939303 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694038 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2597485 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001280.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39229.10.14_5-S | - | - |
