Assay Details
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024229.2 | NP_077191.2 | 2 - 3 | 216 | 76 | |
GenBank mRNA | AK140458.1 | - | 2 - 3 | 256 | 76 | |
AK144544.1 | - | 2 - 3 | 263 | 76 | ||
AK166319.1 | - | 2 - 3 | 245 | 76 | ||
BC003473.1 | - | 2 - 3 | 222 | 2650077 | 76 | |
BC008276.1 | - | 2 - 3 | 216 | 3707732 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
