Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271898.1 | NP_001258827.1 | 1 - 2 | 448 | 75 | |
| NM_015729.3 | NP_056544.2 | 1 - 2 | 448 | 75 | ||
| GenBank mRNA | AB034914.1 | - | 1 - 2 | 137 | 75 | |
| AF006688.1 | - | 1 - 2 | 187 | 75 | ||
| AK040566.1 | - | 1 - 2 | 151 | 75 | ||
| AK054446.1 | - | 1 - 2 | 224 | 75 | ||
| AK128941.1 | - | 1 - 2 | 150 | 75 | ||
| BC054727.1 | - | 1 - 2 | 116 | 75 | ||
| BC056448.1 | - | 1 - 2 | 448 | 5704873 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map