Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015729.3 | NP_056544.2 | 2 - 3 | 611 | 70 | |
| GenBank mRNA | AK040566.1 | - | 2 - 3 | 314 | 70 | |
| AK054446.1 | - | 2 - 3 | 387 | 70 | ||
| BC054727.1 | - | 2 - 3 | 279 | 70 | ||
| BC056448.1 | - | 2 - 3 | 611 | 5704873 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox1
Genomic Map